α-Brom-2-fluortoluol ≥98%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Purity: 98%
CAS No.: 446-48-0
Molecular Formula: C7H6BrF
Formula Weight: 189.03
Flash Point: 82°(179°F)
Refractive Index: 1.5520
Odour: Pungent
Boiling Point: 84-85°/15mm
Beilstein Reference: 1099894
UN No.: 3265
MDL No.: MFCD00000324
Formel:
C₇H₆BrF Siedepunkt: 84…85 °C (15 mmHg) Dichte: 1,567 Flammpunkt: 82 °C (179 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00000324 CAS-Nummer: 446-48-0 EINECS: 207-169-1 UN: 3265 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...