Acetophenon ≥99%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
A12727.AP
A12727.0F
A12727.0D
A12727.APEA
31
EUR
A12727.AP
A12727.0F
A12727.0D
Acetophenon ≥99%
Acetophenon
Purity: 99%
CAS No.: 98-86-2
Molecular Formula: C8H8O
Formula Weight: 120.15
Flash Point: 76°(169°F)
Refractive Index: 1.5325
Melting Point: 19-20°
Boiling Point: 201-203°
Merck Reference: 14,73
MDL No.: MFCD00008724
Formel:
C₈H₈O Siedepunkt: 201…203 °C Schmelzpunkt: 19…20 °C Dichte: 1,030 Flammpunkt: 76 °C (169 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00008724 CAS-Nummer: 98-86-2 EINECS: 202-708-7 UN: 3334 ADR: 9, Merck Index: 14,73 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...