2,2,6,6-Tetramethylpiperidin-1-oxyl (TEMPO) ≥98% freies Radikal
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Purity: 98+%
CAS No.: 2564-83-2
Molecular Formula: C9H18NO
Formula Weight: 156.25
Storage Temperature: 2°C to 8°C
Flash Point: 67°(152°F)
Melting Point: 31-40°
Boiling Point: 193° dec.
Beilstein Reference: 1422418
Merck Reference: 14,9140
UN No.: 3263
MDL No.: MFCD00009599
Formel:
C₉H₁₈NO Siedepunkt: 193 °C (Zersetzung) Schmelzpunkt: 31…40 °C Flammpunkt: 67 °C (152 °F) Lagertemperatur: Kühlschrank |
MDL:
MFCD00009599 CAS-Nummer: 2564-83-2 EINECS: 219-888-8 UN: 3263 ADR: 8,III Merck Index: 14,9140 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...