(3-Fluorphenyl)acetonitril ≥99%

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: 3-Fluorbenzylcyanid

A12737.06 A12737.14
A12737.06EA 37.6 EUR
A12737.06 A12737.14
(3-Fluorphenyl)acetonitril ≥99%
(3-Fluorphenyl)acetonitril

Purity: 99% CAS No.: 501-00-8 Molecular Formula: C8H6FN Formula Weight: 135.14 Flash Point: 110°(230°F) Refractive Index: 1.5020 Boiling Point: 230° Beilstein Reference: 1861071 UN No.: 3276 MDL No.: MFCD00001907

Formel: C₈H₆FN
Molecular Weight: 135.14 g/mol
Siedepunkt: 230 °C
Dichte: 1.156
Flammpunkt: >110 °C (230 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00001907
CAS-Nummer: 501-00-8
EINECS: 207-918-2
UN: 3276
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR