4-Methoxy-β-nitrostyrol ≥98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
trans-4-Methoxy-β-nitrostyrol
A12745.06
A12745.14
A12745.22
A12745.06EA
76.9
EUR
A12745.06
A12745.14
A12745.22
4-Methoxy-β-nitrostyrol ≥98%
4-Methoxy-β-nitrostyrol
Purity: 98%
CAS No.: 5576-97-6
Molecular Formula: C9H9NO3
Formula Weight: 179.18
Storage Temperature: 2°C to 8°C
Melting Point: 86-88°
Beilstein Reference: 2047935
MDL No.: MFCD00024826
Formel:
C₉H₉NO₃ Schmelzpunkt: 86…88 °C |
MDL:
MFCD00024826 CAS-Nummer: 3179-10-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...