Benzimidazol ≥99%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 1H-Benzimidazol

A12763.0B A12763.18 A12763.30
A12763.0BEA 254 EUR
A12763.0B A12763.18 A12763.30
Benzimidazol ≥99%
Benzimidazol

Purity: 99% CAS No.: 51-17-2 Molecular Formula: C7H6N2 Formula Weight: 118.14 Melting Point: 170-174° Boiling Point: 360° Merck Reference: 14,1081 MDL No.: MFCD00005585

Formel: C₇H₆N₂
Siedepunkt: 360 °C
Schmelzpunkt: 170…174 °C
Dichte: 1,23 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00005585
CAS-Nummer: 51-17-2
EINECS: 200-081-4
Merck Index: 14,1081

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR