Benzimidazol ≥99%
Lieferant: Thermo Fisher Scientific
Achtung
|
Synonyme:
1H-Benzimidazol
A12763.0B
A12763.18
A12763.30
A12763.0BEA
254
EUR
A12763.0B
A12763.18
A12763.30
Benzimidazol ≥99%
Benzimidazol
Purity: 99%
CAS No.: 51-17-2
Molecular Formula: C7H6N2
Formula Weight: 118.14
Melting Point: 170-174°
Boiling Point: 360°
Merck Reference: 14,1081
MDL No.: MFCD00005585
Formel:
C₇H₆N₂ Siedepunkt: 360 °C Schmelzpunkt: 170…174 °C Dichte: 1,23 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00005585 CAS-Nummer: 51-17-2 EINECS: 200-081-4 Merck Index: 14,1081 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...