1,2-Dibromethan ≥99%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
A12766.0B
A12766.30
A12766.0BEA
126
EUR
A12766.0B
A12766.30
1,2-Dibromethan ≥99%
1,2-Dibromethan
Purity: 99%
CAS No.: 106-93-4
Molecular Formula: C2H4Br2
Formula Weight: 187.87
Flash Point: None
Refractive Index: 1.5385
Melting Point: 9-10°
Boiling Point: 132°
Merck Reference: 14,3796
UN No.: 1605
MDL No.: MFCD00000233
Formel:
BrH₂CCH₂Br Siedepunkt: 132 °C Schmelzpunkt: 9…10 °C Dichte: 2,179 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00000233 CAS-Nummer: 106-93-4 EINECS: 203-444-5 UN: 1605 ADR: 6.1,I Merck Index: 14,3796 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...