1,2-Dichlorethan ≥99%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Purity: 99+%
CAS No.: 107-06-2
Molecular Formula: C2H4Cl2
Formula Weight: 98.96
Flash Point: 15°(59°F)
Refractive Index: 1.4448
Melting Point: -35°
Boiling Point: 83°
Beilstein Reference: 605264
Merck Reference: 14,3797
UN No.: 1184
MDL No.: MFCD00000963
Formel:
ClH₂CCH₂Cl Siedepunkt: 83 °C Schmelzpunkt: –35 °C Dichte: 1,256 Flammpunkt: 15 °C (59 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00000963 CAS-Nummer: 107-06-2 EINECS: 203-458-1 UN: 1184 ADR: 3,II Merck Index: 14,3797 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...