Thiomersal
Lieferant: PanReac AppliChem
|
Gefahr
|
A1278.0100
A1278.0100EA
1350
EUR
A1278.0100
Thiomersal
Thiomersal
Formel:
2-(C₂H₅HgS)C₆H₄CO₂Na Schmelzpunkt: 232…233 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00013062 CAS-Nummer: 54-64-8 EINECS: 200-210-4 UN: 2025 ADR: 6.1,II Merck Index: 12,09451 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...