Brillantgrün (Malachitgrün G)
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
Ethyl Green, CI 42040, Basic Green 1, N-(4-((4-(Diethylamino)phenyl)(phenyl)methylene)cyclohexa-2,5-dien-1-ylidene)-N-ethylethanaminium hydrogen sulphate, Diamond Green, Malachite Green G, Emerald Green, Astradiamant green GX, Solid Green JO, Brilliant Green hydrogen sulphate
A12801.14
A12801.22
A12801.14EA
20.9
EUR
A12801.14
A12801.22
Brillantgrün (Malachitgrün G)
Brillantgrün (Malachitgrün G)
CAS No.: 633-03-4
Molecular Formula: C27H34N2O4S
Formula Weight: 482.65
Melting Point: 210° dec.
Beilstein Reference: 3901207
Merck Reference: 14,1374
MDL No.: MFCD00011880
Formel:
C₂₇H₃₄N₂O₄S Schmelzpunkt: 210 °C (Zers.) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00011880 CAS-Nummer: 633-03-4 EINECS: 211-190-1 Merck Index: 14,1374 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...