Magnesiumfluorid Hydrat (≥87% MgF₂)
Lieferant: Thermo Fisher Scientific
A12830.0E
A12830.36
A12830.0EEA
131
EUR
A12830.0E
A12830.36
Magnesiumfluorid Hydrat (≥87% MgF₂)
Magnesiumfluorid Hydrat
CAS No.: 7783-40-6
Molecular Formula: MgF2.xH2O
Formula Weight: 62.30(anhy)
Odour: Odourless
Merck Reference: 14,5665
MDL No.: MFCD00011108
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...