2,5-Dimethylfuran ≥98%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
A12833.14
A12833.22
A12833.36
A12833.14EA
49.2
EUR
A12833.14
A12833.22
A12833.36
2,5-Dimethylfuran ≥98%
2,5-Dimethylfuran
Purity: 98+%
CAS No.: 625-86-5
Molecular Formula: C6H8O
Formula Weight: 96.13
Flash Point: 1°(33°F)
Refractive Index: 1.4410
Melting Point: -62°
Boiling Point: 92-93°
Beilstein Reference: 106449
UN No.: 3271
MDL No.: MFCD00003250
Formel:
C₆H₈O Molecular Weight: 96.13 g/mol Siedepunkt: 92…93 °C Schmelzpunkt: –62 °C Dichte: 0.903 Flammpunkt: 1 °C (33 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00003250 CAS-Nummer: 625-86-5 EINECS: 210-914-3 UN: 3271 ADR: 3 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...