2-(4-Chlorphenylthio)benzaldehyd ≥98%
Lieferant: Thermo Fisher Scientific
A12852.06
A12852.14
A12852.06EA
52.8
EUR
A12852.06
A12852.14
2-(4-Chlorphenylthio)benzaldehyd ≥98%
2-(4-Chlorophenylthio)benzaldehyde
Purity: 98%
CAS No.: 107572-07-6
Molecular Formula: C13H9ClOS
Formula Weight: 248.72
Melting Point: 71-74°
Boiling Point: 102-104°/14mm
MDL No.: MFCD00051766
Formel:
C₁₃H₉ClOS Siedepunkt: 102…104 °C (14 mmHg) Schmelzpunkt: 71…74 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00051766 CAS-Nummer: 107572-07-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...