2,4-Dimethyl-3-pentanon ≥98%
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
2,4-Dimethylpentan-3-one, Diisopropyl ketone, Isobutyrone
A12875.22
A12875.22EA
32.4
EUR
A12875.22
2,4-Dimethyl-3-pentanon ≥98%
2,4-Dimethyl-3-pentanon
Purity: 98%
CAS No.: 565-80-0
Molecular Formula: C7H14O
Formula Weight: 114.19
Flash Point: 15°(59°F)
Refractive Index: 1.3990
Melting Point: -69°
Boiling Point: 124-125°
Beilstein Reference: 773782
UN No.: 1224
MDL No.: MFCD00008918
Formel:
C₇H₁₄O Molecular Weight: 114,19 g/mol Siedepunkt: 124…125 °C Schmelzpunkt: –69 °C Dichte: 0,803 Flammpunkt: 15 °C (59 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00008918 CAS-Nummer: 565-80-0 EINECS: 209-294-7 UN: 1224 ADR: 3,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...