Pluronic® F-68, BioChemica
Lieferant: PanReac AppliChem
A1288.0500
A1288.0500EA
113
EUR
A1288.0500
Pluronic® F-68, BioChemica
Kolliphor® P 188 (Lutrol® F68, Pluronic® F-68, Synperonic® F68)
Formel:
HO(C₂H₄O)a(C₃H₆O)b(C₂H₄O)a Lagertemperatur: Raumtemperatur |
MDL:
MFCD00082049 CAS-Nummer: 9003-11-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...