Tris[4-(trifluoromethyl)phenyl]phosphine 98%

Lieferant: Thermo Scientific
Achtung

Synonyme: Tris(4-trifluoromethylphenyl)phosphine, Tris(4-(trifluoromethyl)phenyl)phosphine

A12885.03
A12885.03EA 142 EUR
A12885.03
Tris[4-(trifluoromethyl)phenyl]phosphine 98%
Tris[4-(trifluoromethyl)phenyl]phosphine

Purity: 98% CAS No.: 13406-29-6 Molecular Formula: C21H12F9P Formula Weight: 466.29 Melting Point: 72-75° MDL No.: MFCD00058883

Formel: C₂₁H₁₂F₉P
Molecular Weight: 466,28 g/mol
Schmelzpunkt: 72…75 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00058883
CAS-Nummer: 13406-29-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR