Zinkacetat Dihydrat ≥97%

Lieferant: Thermo Fisher Scientific
Achtung

A12909.0B A12909.0I A12909.22 A12909.36 A12909.A4
A12909.0BEA 76.7 EUR
A12909.0B A12909.0I A12909.22 A12909.36 A12909.A4
Zinkacetat Dihydrat ≥97%
Zinkacetat Dihydrat

Purity: 97+% CAS No.: 5970-45-6 Molecular Formula: Zn(OOCCH3)2.2H2O Formula Weight: 219.50 (183.47anhy) Odour: Weak, characteristic pH: 6-7 Melting Point: 237° Beilstein Reference: 3732513 Merck Reference: 14,10128 UN No.: 3077 MDL No.: MFCD00066961

Formel: (H₃CCOO)₂Zn·2H₂O
Schmelzpunkt: 237 °C
Dichte: 1,735
Lagertemperatur: Raumtemperatur
MDL: MFCD00066961
CAS-Nummer: 5970-45-6
EINECS: 209-170-2
UN: 3077
ADR: 9,III
Merck Index: 14,10128

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR