9-Fluorenon ≥98%

Lieferant: Thermo Fisher Scientific

Synonyme: 9H-Fluoren-9-one

A12910.0E A12910.22 A12910.36
A12910.0EEA 537 EUR
A12910.0E A12910.22 A12910.36
9-Fluorenon ≥98%
9-Fluorenon

Purity: 98+% CAS No.: 486-25-9 Molecular Formula: C13H8O Formula Weight: 180.21 Flash Point: 163°(325°F) Physical Form: Cryst powder or flakes Density: 0.9 Appearance: Yellow Melting Point: 82-85° Boiling Point: 342° Beilstein Reference: 1636531 MDL No.: MFCD00001141

Formel: C₁₃H₈O
Molecular Weight: 180.21 g/mol
Siedepunkt: 342 °C
Schmelzpunkt: 82…85 °C
Dichte: 0.9
Flammpunkt: 163 °C (325 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00001141
CAS-Nummer: 486-25-9
EINECS: 207-630-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR