2,6-Dibrom-4-nitrophenol ≥98%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 1,3-Dibromo-2-hydroxy-5-nitrobenzene

A12916.14 A12916.22 A12916.30
A12916.14EA 45.8 EUR
A12916.14 A12916.22 A12916.30
2,6-Dibrom-4-nitrophenol ≥98%
2,6-Dibrom-4-nitrophenol

Purity: 98% CAS No.: 99-28-5 Molecular Formula: C6H3Br2NO3 Formula Weight: 296.90 Melting Point: ca 145° dec. Beilstein Reference: 1245050

Formel: C₆H₃Br₂NO₃
Molecular Weight: 296.9 g/mol
Schmelzpunkt: ∼ 145 °C (Zers.)
Lagertemperatur: Raumtemperatur
MDL: MFCD00007334
CAS-Nummer: 99-28-5
EINECS: 202-744-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR