6-Nitroindol ≥97%
Lieferant: Thermo Scientific
Synonyme:
6-Nitro-1H-indole
A12927.03
A12927.03EA
60.4
EUR
A12927.03
6-Nitroindol ≥97%
6-Nitroindol
Purity: 97%
CAS No.: 4769-96-4
Molecular Formula: C8H6N2O2
Formula Weight: 162.15
Melting Point: 141-145°
Beilstein Reference: 135977
Formel:
C₈H₆N₂O₂ Molecular Weight: 162,15 g/mol Schmelzpunkt: 141…145 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00051497 CAS-Nummer: 4769-96-4 EINECS: 225-311-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...