Anisol ≥99%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Purity: 99%
CAS No.: 100-66-3
Molecular Formula: C7H8O
Formula Weight: 108.14
Flash Point: 43°(109°F)
Refractive Index: 1.5170
Odour: Ether-like
Melting Point: -37°
Boiling Point: 153-154°
Beilstein Reference: 506892
Merck Reference: 14,669
UN No.: 2222
MDL No.: MFCD00008354
Formel:
C₇H₈O Siedepunkt: 153…154 °C Schmelzpunkt: –37 °C Dichte: 0,994 Flammpunkt: 43 °C (109 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00008354 CAS-Nummer: 100-66-3 EINECS: 202-876-1 UN: 2222 ADR: 3,III Merck Index: 14,669 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...