3-Nitro-o-kresol 96%

Lieferant: Thermo Scientific
Gefahr

Synonyme: 1-Hydroxy-2-methyl-3-nitrobenzol, 2-Methyl-3-nitrophenol, 2-Hydroxy-6-nitrotoluol

A13002.14
A13002.14EA 337 EUR
A13002.14
3-Nitro-o-kresol 96%
3-Nitro-o-kresol

Purity: 96% CAS No.: 5460-31-1 Molecular Formula: C7H7NO3 Formula Weight: 153.14 Odour: Phenol-like Melting Point: 148-152° Beilstein Reference: 1946057 UN No.: 2446 MDL No.: MFCD00007241

Formel: C₇H₇NO₃
Molecular Weight: 153,14 g/mol
Schmelzpunkt: 148…152 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00007241
CAS-Nummer: 5460-31-1
EINECS: 226-739-0
UN: 2446
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR