2,6-Difluoranilin ≥98%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 2,6-Difluorphenylamin

A13020.06 A13020.14 A13020.22
A13020.06EA 34 EUR
A13020.06 A13020.14 A13020.22
2,6-Difluoranilin ≥98%
2,6-Difluoranilin

Purity: 98% CAS No.: 5509-65-9 Molecular Formula: C6H5F2N Formula Weight: 129.11 Flash Point: 51°(124°F) Refractive Index: 1.5080 Odour: Characteristic Boiling Point: 152-154° Beilstein Reference: 2802697 UN No.: 1992 MDL No.: MFCD00007655

Formel: C₆H₅F₂N
Molecular Weight: 129.11 g/mol
Siedepunkt: 152…154 °C
Dichte: 1.275
Flammpunkt: 51 °C (124 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00007655
CAS-Nummer: 5509-65-9
EINECS: 226-853-0
UN: 1992
ADR: 3 III

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR