3-Phenyl-1-propanol ≥99%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
A13022.0B
A13022.0I
A13022.30
A13022.0BEA
84.4
EUR
A13022.0B
A13022.0I
A13022.30
3-Phenyl-1-propanol ≥99%
3-Phenyl-1-propanol
Purity: 99%
CAS No.: 122-97-4
Molecular Formula: C9H12O
Formula Weight: 136.19
Flash Point: 109°(228°F)
Refractive Index: 1.5260
Melting Point: -18°
Boiling Point: 235°
Beilstein Reference: 1857542
MDL No.: MFCD00002950
Formel:
(C₆H₅)CH₂CH₂CH₂OH Siedepunkt: 235 °C Schmelzpunkt: –18 °C Dichte: 1,004 Flammpunkt: 109 °C (228 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00002950 CAS-Nummer: 122-97-4 EINECS: 204-587-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...