2-Aminopyrazin 99%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: Aminopyrazin, Pyrazin-2-amin

A13052.06 A13052.14 A13052.18
A13052.06EA 39 EUR
A13052.06 A13052.14 A13052.18
2-Aminopyrazin 99%
2-Aminopyrazine

Purity: 99% CAS No.: 5049-61-6 Molecular Formula: C4H5N3 Formula Weight: 95.11 Melting Point: 118-121° Beilstein Reference: 107025

Formel: C₄H₅N₃
Siedepunkt: 203 °C (1013 hPa)
Schmelzpunkt: 118…121 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00006137
CAS-Nummer: 5049-61-6
EINECS: 225-748-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR