Ethyl-3-phenylpropionat ≥98%
Lieferant: Thermo Scientific
Synonyme:
3-Phenylpropionsäureethylester, Hydrozimtsäureethylester, Ethyldihydrocinnamat
A13055.18
A13055.18EA
42.6
EUR
A13055.18
Ethyl-3-phenylpropionat ≥98%
Ethyl-3-phenylpropionat
Purity: 98+%
CAS No.: 2021-28-5
Molecular Formula: C6H5CH2CH2CO2CH2CH3
Formula Weight: 178.23
Flash Point: 107°(224°F)
Refractive Index: 1.4940
Boiling Point: 118°/3mm
Beilstein Reference: 1910432
MDL No.: MFCD00009206
Formel:
C₁₁H₁₄O₂ Molecular Weight: 178,23 g/mol Siedepunkt: 118 °C (3 mmHg) Dichte: 1,016 Flammpunkt: 107 °C (224 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00009206 CAS-Nummer: 2021-28-5 EINECS: 217-966-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...