2-(1-Piperidinyl)anilin ≥98%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
2-(1-Piperidino)aniline
A13073.03
A13073.06
A13073.14
A13073.03EA
22.9
EUR
A13073.03
A13073.06
A13073.14
2-(1-Piperidinyl)anilin ≥98%
2-(1-Piperidinyl)anilin
Purity: 98%
CAS No.: 39643-31-7
Molecular Formula: C11H16N2
Formula Weight: 176.26
Melting Point: 44-47°
Boiling Point: 96-98°/1mm
UN No.: 2811
MDL No.: MFCD00047467
Formel:
C₁₁H₁₆N₂ Siedepunkt: 96…98 °C (1 mmHg) Schmelzpunkt: 44…47 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00047467 CAS-Nummer: 39643-31-7 UN: 2811 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...