2,6-Di-tert-butyl-p-benzochinon ≥98%

Lieferant: Thermo Fisher Scientific
Achtung

A13091.06 A13091.14
A13091.06EA 72.8 EUR
A13091.06 A13091.14
2,6-Di-tert-butyl-p-benzochinon ≥98%
2,6-Di-tert-butyl-p-benzochinon

Purity: 98+% CAS No.: 719-22-2 Molecular Formula: C14H20O2 Formula Weight: 220.31 Flash Point: 110°(230°F) Melting Point: 65-69° Beilstein Reference: 1911512 MDL No.: MFCD00001601

Formel: C₁₄H₂₀O₂
Schmelzpunkt: 65…69 °C
Flammpunkt: >110 °C (230 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00001601
CAS-Nummer: 719-22-2
EINECS: 211-946-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR