1-Pentanol ≥98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
1-Pentylalkohol, n-Amylalkohol, Butylcarbinol, 1-Hydroxypentan
A13093.0D
A13093.0F
A13093.AP
A13093.0DEA
229
EUR
A13093.0D
A13093.0F
A13093.AP
1-Pentanol ≥98%
1-Pentanol
Purity: 98+%
CAS No.: 71-41-0
Molecular Formula: C5H12O
Formula Weight: 88.15
Flash Point: 49°(120°F)
Refractive Index: 1.4103
Odour: Mild
Melting Point: -79°
Boiling Point: 136-138°
Beilstein Reference: 1730975
Merck Reference: 14,7118
UN No.: 1105
MDL No.: MFCD00002977
Formel:
H₃C(CH₂)₃CH₂OH Molecular Weight: 88.15 g/mol Siedepunkt: 136…138 °C Schmelzpunkt: –79 °C Dichte: 0.812 Flammpunkt: 49 °C (120 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00002977 CAS-Nummer: 71-41-0 EINECS: 200-752-1 UN: 1105 ADR: 3 III Merck Index: 13,07195 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...