Dimethylcarbonat ≥99%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
Kohlensäuredimethylester, Methylcarbonat
A13104.0E
A13104.22
A13104.36
A13104.0EEA
111
EUR
A13104.0E
A13104.22
A13104.36
Dimethylcarbonat ≥99%
Dimethylcarbonat
Purity: 99%
CAS No.: 616-38-6
Molecular Formula: C3H6O3
Formula Weight: 90.08
Flash Point: 16°(61°F)
Physical Form: Liquid
Density: 1.070
Refractive Index: 1.3680
Sensitivity: Moisture Sensitive
Appearance: Colourless
Odour: Pleasant
Melting Point: ca 4°
Boiling Point: 90°
Solubility: Insoluble in water. Miscible with alcohol, ether
Beilstein Reference: 635821
Merck Reference: 14,3241
UN No.: 1161
MDL No.: MFCD00008420
Formel:
C₃H₆O₃ Siedepunkt: 90 °C Schmelzpunkt: ∼ 4 °C Dichte: 1,070 Flammpunkt: 16 °C (61 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00008420 CAS-Nummer: 616-38-6 EINECS: 210-478-4 UN: 1161 ADR: 3,II Merck Index: 14,3241 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...