Chlordimethylsilan 97%, Stäbe
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Purity: 97%
CAS No.: 1066-35-9
Molecular Formula: C2H7ClSi
Formula Weight: 94.62
Storage Temperature: 2°C to 8°C
Flash Point: -28°(-18°F)
Refractive Index: 1.3830
Odour: Acrid
Melting Point: -111°
Boiling Point: 34-36°
Beilstein Reference: 1633434
UN No.: 2924
MDL No.: MFCD00000495
Formel:
C₂H₇ClSi Siedepunkt: 34…36 °C Schmelzpunkt: –111 °C Dichte: 0,869 Flammpunkt: –28 °C (–18 °F) Lagertemperatur: Kühlschrank |
MDL:
MFCD00000495 CAS-Nummer: 1066-35-9 EINECS: 213-912-0 UN: 2924 ADR: 3,I REACH: 01-2119486782-25 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...