Fmoc-OSu (N-(9-Fluorenylmethoxycarbonyloxy)succinimid) ≥98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
Fmoc-OSu, N-(9-Fluorenylmethoxycarbonyloxy)succinimide
A13143.03
A13143.06
A13143.14
A13143.03EA
21.1
EUR
A13143.03
A13143.06
A13143.14
Fmoc-OSu (N-(9-Fluorenylmethoxycarbonyloxy)succinimid) ≥98%
Fmoc-OSu (N-(9-Fluorenylmethoxycarbonyloxy)succinimid)
Purity: 98%
CAS No.: 82911-69-1
Molecular Formula: C19H15NO5
Formula Weight: 337.34
Storage Temperature: 2°C to 8°C
Melting Point: 150-153°
Beilstein Reference: 3569540
UN No.: 3077
MDL No.: MFCD00010733
Formel:
C₁₉H₁₅NO₅ Schmelzpunkt: 150…153 °C |
MDL:
MFCD00010733 CAS-Nummer: 82911-69-1 EINECS: 433-520-5 ADR: 9,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...