6-Chlor-2-hydroxypyridin ≥98%
Lieferant: Thermo Fisher Scientific
A13164.14
A13164.22
A13164.14EA
61.5
EUR
A13164.14
A13164.22
6-Chlor-2-hydroxypyridin ≥98%
Purity: 98%
CAS No.: 16879-02-0
Molecular Formula: C5H4ClNO
Formula Weight: 129.55
Flash Point: 163°(325°F)
Melting Point: 129-131°
Beilstein Reference: 108665
MDL No.: MFCD00006278
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...