Natriumacetat, wasserfrei 99%

Lieferant: Thermo Fisher Scientific
Achtung

A13184.0B A13184.0I A13184.30
A13184.0BEA 42 EUR
A13184.0B A13184.0I A13184.30
Natriumacetat, wasserfrei 99%
Natriumacetat

Purity: 99% CAS No.: 127-09-3 Molecular Formula: C2H3NaO2 Formula Weight: 82.03 pH: 7,5-9 Melting Point: 324° Beilstein Reference: 3595639 Merck Reference: 14,8571 MDL No.: MFCD00012459

Formel: H₃CCOONa
Siedepunkt: 881,4 °C (1013 hPa)
Schmelzpunkt: 324 °C
Dichte: 1,528
Lagertemperatur: Raumtemperatur
MDL: MFCD00012459
CAS-Nummer: 127-09-3
EINECS: 204-823-8
REACH: 01-2119485123-42
Merck Index: 14,8571

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR