1,2-Bis(2-aminophenoxy)ethane-N,N,N',N'-tetraacetic acid 97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
A13190.03
A13190.06
A13190.14
A13190.03EA
133
EUR
A13190.03
A13190.06
A13190.14
1,2-Bis(2-aminophenoxy)ethane-N,N,N',N'-tetraacetic acid 97%
1,2-Bis(2-aminophenoxy)ethane-N,N,N',N'-tetraacetic acid
Purity: 97%
CAS No.: 85233-19-8
Molecular Formula: C22H24N2O10
Formula Weight: 476.44
Storage Temperature: 2°C to 8°C
Melting Point: ca 174° dec.
MDL No.: MFCD00036255
Formel:
C₂₂H₂₄N₂O₁₀ Lagertemperatur: Kühlschrank |
MDL:
MFCD00036255 CAS-Nummer: 85233-19-8 Merck Index: 14,957 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...