5-Methyl-2-furaldehyd ≥98%

Lieferant: Thermo Fisher Scientific
Achtung

A13264.22 A13264.14
A13264.22EA 100 EUR
A13264.22 A13264.14
5-Methyl-2-furaldehyd ≥98%
5-Methyl-2-furaldehyd

Purity: 98% CAS No.: 620-02-0 Molecular Formula: C6H6O2 Formula Weight: 110.11 Flash Point: 72°(161°F) Refractive Index: 1.5310 Boiling Point: 186-188° Beilstein Reference: 106895 MDL No.: MFCD00003232

Formel: C₆H₆O₂
Siedepunkt: 186…188 °C
Dichte: 1,106
Flammpunkt: 72 °C (161 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00003232
CAS-Nummer: 620-02-0
EINECS: 210-622-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR