Benzyltriethylammoniumchlorid ≥99%

Lieferant: Thermo Fisher Scientific
Achtung

A13268.22 A13268.36
A13268.22EA 25.8 EUR
A13268.22 A13268.36
Benzyltriethylammoniumchlorid ≥99%
Benzyltriethylammoniumchlorid

Purity: 99% CAS No.: 56-37-1 Molecular Formula: (CH3CH2)3N(CH2C6H5)Cl Formula Weight: 227.78 Melting Point: ca 190° dec. Beilstein Reference: 3574984

Formel: C₁₃H₂₂ClN
Flammpunkt: >100 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00011824
CAS-Nummer: 56-37-1
EINECS: 200-270-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR