Bromacetaldehyddiethylacetal ≥97%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Purity: 97%
CAS No.: 2032-35-1
Molecular Formula: C6H13BrO2
Formula Weight: 197.08
Storage Temperature: 2°C to 8°C
Flash Point: 61°(142°F)
Refractive Index: 1.4390
Boiling Point: 66-67°/18mm
Beilstein Reference: 635754
UN No.: 2810
MDL No.: MFCD00000214
Formel:
BrH₂CCH(OCH₂CH₃)₂ Siedepunkt: 66…67 °C (18 mmHg) Dichte: 1,270 Flammpunkt: 61 °C (142 °F) Lagertemperatur: Kühlschrank |
MDL:
MFCD00000214 CAS-Nummer: 2032-35-1 EINECS: 217-989-1 UN: 2810 ADR: 6.1,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...