3-Picolylchlorid Hydrochlorid ≥97%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
A13279.06
A13279.14
A13279.06EA
30.9
EUR
A13279.06
A13279.14
3-Picolylchlorid Hydrochlorid ≥97%
3-Picolylchlorid Hydrochlorid
Purity: 97%
CAS No.: 6959-48-4
Molecular Formula: C6H6ClN.HCl
Formula Weight: 164.04
pH: 3
Melting Point: 136-142°
Beilstein Reference: 3689156
UN No.: 3261
MDL No.: MFCD00012818
Formel:
C₆H₇Cl₂N Schmelzpunkt: 136…142 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00012818 CAS-Nummer: 6959-48-4 EINECS: 230-150-4 UN: 3261 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...