6-Chlornicotinonitril ≥98%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
2-Chlor-5-pyridincarbonitril, 6-Chlornicotinsäurenitril, 2-Chlor-5-cyanpyridin, 2-Chlor-5-pyridincarbonsäurenitril
A13291.06
A13291.03
A13291.14
A13291.06EA
60.5
EUR
A13291.06
A13291.03
A13291.14
6-Chlornicotinonitril ≥98%
6-Chlornicotinonitril
Purity: 98%
CAS No.: 33252-28-7
Molecular Formula: C6H3ClN2
Formula Weight: 138.56
Melting Point: 116-119°
Boiling Point: 105-107°/1mm
UN No.: 3439
MDL No.: MFCD00084941
Formel:
C₆H₃ClN₂ Siedepunkt: 105…107 °C (1 mmHg) Schmelzpunkt: 116…119 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00084941 CAS-Nummer: 33252-28-7 UN: 3439 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...