di-Phosphorpentoxid ≥98%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
A13348.0E
A13348.0EEA
114
EUR
A13348.0E
di-Phosphorpentoxid ≥98%
di-Phosphorpentoxid
Purity: 98%
CAS No.: 1314-56-3
Molecular Formula: P2O5
Formula Weight: 141.94
Odour: Weak
Melting Point: 340°
Boiling Point: 360° subl.
Merck Reference: 14,7355
UN No.: 1807
MDL No.: MFCD00011440
Formel:
P₂O₅ Siedepunkt: 360 °C (Subl.) Schmelzpunkt: 340 °C Dichte: 2,39 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00011440 CAS-Nummer: 1314-56-3 EINECS: 215-236-1 UN: 1807 ADR: 8,II Merck Index: 14,7355 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...