(±)-4-Hydroxy-3-methoxymandelsäure ≥98%

Lieferant: Thermo Fisher Scientific
Achtung

A13349.14 A13349.06 A13349.03
A13349.14EA 782 EUR
A13349.14 A13349.06 A13349.03
(±)-4-Hydroxy-3-methoxymandelsäure ≥98%
(±)-4-Hydroxy-3-methoxymandelsäure

Purity: 98% CAS No.: 2394-20-9 Molecular Formula: C9H10O5 Formula Weight: 198.17 Melting Point: 133-134° dec. MDL No.: MFCD00004235

Formel: C₉H₁₀O₅
Siedepunkt: 421 °C (1013 hPa)
Dichte: 1,44 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00004235
CAS-Nummer: 55-10-7
EINECS: 200-224-0
Merck Index: 12,10070

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR