3,4-Dibromtoluol ≥98%
Lieferant: Thermo Fisher Scientific
Synonyme:
1,2-Dibromo-4-methylbenzene
A13351.06
A13351.14
A13351.22
A13351.06EA
24.7
EUR
A13351.06
A13351.14
A13351.22
3,4-Dibromtoluol ≥98%
3,4-Dibromtoluol
Purity: 98+%
CAS No.: 60956-23-2
Molecular Formula: C7H6Br2
Formula Weight: 249.94
Flash Point: 116°(241°F)
Refractive Index: 1.6000
Melting Point: -10°
Boiling Point: 251°
Beilstein Reference: 1931706
MDL No.: MFCD00079744
Formel:
C₇H₆Br₂ |
MDL:
MFCD00079744 CAS-Nummer: 60956-23-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...