Dichlon ≥98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
2,3-Dichlor-1,4-naphthochinon, 2,3-Chlorbenzo(e)cyclohex-2-en-1,4-dion
A13378.14
A13378.22
A13378.36
A13378.14EA
20.9
EUR
A13378.14
A13378.22
A13378.36
Dichlon ≥98%
Dichlon
Purity: 98%
CAS No.: 117-80-6
Molecular Formula: C10H4Cl2O2
Formula Weight: 227.05
Melting Point: 193-196° subl.
Boiling Point: 275°/2mm
Merck Reference: 14,3045
UN No.: 2811
MDL No.: MFCD00001677
Formel:
C₁₀H₄Cl₂O₂ Molecular Weight: 227.05 g/mol Siedepunkt: 275 °C (2 mmHg) Schmelzpunkt: 193…196 °C (Subl.) Dichte: 1.44 g/cm³ (16 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00001677 CAS-Nummer: 117-80-6 EINECS: 204-210-5 UN: 2811 ADR: 6.1 III Merck Index: 13,03071 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...