L(+)-Histidin
Lieferant: PanReac AppliChem
A1341.1000
A1341.1000EA
328
EUR
A1341.1000
L(+)-Histidin
L(+)-Histidin
Formel:
C₆H₉N₃O₂ Schmelzpunkt: 272…273 °C Dichte: 1,44 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00064315 CAS-Nummer: 71-00-1 EINECS: 200-745-3 Merck Index: 13,04741 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...