Cyclobutylamin 98%

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: Aminocyclobutan

A13423.03 A13423.06 A13423.14
A13423.03EA 56.1 EUR
A13423.03 A13423.06 A13423.14
Cyclobutylamin 98%
Cyclobutylamin

Purity: 98% CAS No.: 2516-34-9 Molecular Formula: C4H9N Formula Weight: 71.12 Flash Point: -4°(24°F) Refractive Index: 1.4376 Boiling Point: 82-83° Beilstein Reference: 2069297 UN No.: 2733 MDL No.: MFCD00001328

Formel: C₄H₉N
Molecular Weight: 71.12 g/mol
Siedepunkt: 82…83 °C
Dichte: 0.836
Flammpunkt: –4 °C (24 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00001328
CAS-Nummer: 2516-34-9
EINECS: 219-736-0
UN: 2733
ADR: 3 II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR