(±)-4-Methyl-2-pentanol ≥99%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: (±)-Methylisobutylcarbinol, Isobutylmethylcarbinol

A13435.AP A13435.0F
A13435.APEA 23.2 EUR
A13435.AP A13435.0F
(±)-4-Methyl-2-pentanol ≥99%
(±)-4-Methyl-2-pentanol

Purity: 99% CAS No.: 108-11-2 Molecular Formula: C6H14O Formula Weight: 102.18 Flash Point: 41°(105°F) Refractive Index: 1.4110 Odour: Slightly sweetish Melting Point: -90° Boiling Point: 131-133° Beilstein Reference: 1098268 UN No.: 2053 MDL No.: MFCD00004550

Formel: C₆H₁₄O
Molecular Weight: 102.18 g/mol
Siedepunkt: 131…133 °C
Schmelzpunkt: –90 °C
Dichte: 0.806
Flammpunkt: 41 °C (105 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00004550
CAS-Nummer: 108-11-2
EINECS: 203-551-7
UN: 2053
ADR: 3 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR