L-α-Phenylalanin
Lieferant: PanReac AppliChem
Synonyme:
H-Phe-OH, H-L-Phe-OH, L-α-Phenylalanin, 3-Phenyl-L(-)-alanin, 3-Phenyl-L-alanin, (S)-(-)-2-Amino-3-phenylpropansäure
A1344.1000
A1344.1000EA
532
EUR
A1344.1000
L-α-Phenylalanin
L-α-Phenylalanin
Formel:
C₉H₁₁NO₂ Molecular Weight: 165.19 g/mol Siedepunkt: 313 Schmelzpunkt: 173 Dichte: 0.754 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00064227 CAS-Nummer: 63-91-2 EINECS: 200-568-1 Merck Index: 14,07271 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...