1,4-Diphenylbutadiin ≥99%

Lieferant: Thermo Fisher Scientific
Achtung

A13471.14
A13471.14EA 323 EUR
A13471.14
1,4-Diphenylbutadiin ≥99%
1,4-Diphenylbutadiin

Purity: 99% CAS No.: 886-66-8 Molecular Formula: C6H5CC≡CC≡CC6H5 Formula Weight: 202.26 Melting Point: 86-87° Beilstein Reference: 1910105

Formel: (C₆H₅)CCCC(C₆H₅)
Siedepunkt: 338 °C (1013 hPa)
Schmelzpunkt: 87 °C
Dichte: 1,163 g/cm³ (20 °C)
MDL: MFCD00004787
CAS-Nummer: 886-66-8
EINECS: 212-953-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR